Salpetersäure 65 - 70% ≥99,9999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

10984.AC 10984.AK 10984.AP
10984.ACEA 22.1 EUR
10984.AC 10984.AK 10984.AP
Salpetersäure 65 - 70% ≥99,9999% (Metall-Basis)
Salpetersäure

Purity: 99.9999% CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colourless to pale yellow Odour: Pungent Boiling Point: 121° Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349

Formel: HNO₃
Siedepunkt: 121 °C
Schmelzpunkt: –42 °C
Dichte: 1,41
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 14,6577

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR