Cäsiumiodid 99,999 Metals basis

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cesiumiodide

10992.14 10992.22
10992.14EA 114 EUR
10992.14 10992.22
Cäsiumiodid 99,999 Metals basis
Cäsiumiodid

Purity: 99.999% CAS No.: 7789-17-5 Molecular Formula: CsI Formula Weight: 259.81 Physical Form: Crystalline Refractive Index: 1.7876 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 626° Boiling Point: 1280° Merck Reference: 14,2014 MDL No.: MFCD00010962

Formel: CsI
Molecular Weight: 259.81 g/mol
Siedepunkt: 1280 °C
Schmelzpunkt: 626 °C
Dichte: 3.21 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010962
CAS-Nummer: 7789-17-5
EINECS: 232-145-2
Merck Index: 14,2014

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR