Monitor, UVis-920
Lieferant: Cytiva
11-0007-54
11-0007-54EA 7700 EUR
11-0007-54
Monitor, UVis-920
Chromatographie-Detektoren
The monitor UVis-920 is a high-precision, multipurpose, standalone UV monitor. Use of different filters allows the monitor to cover wavelengths from 200 to 700 nm, without changing the lamp.
  • High precision UV/Vis absorbance detection over a wide flow range
  • Easy to use, change of wavelength by filter push-button mechanism without changing the lamp
  • Minimised band broadening and maximised signal-to-noise ratio
  • Minimised footprint