Affinity chromatography columns, His GraviTrap™
Lieferant: Cytiva
11-0033-99
11-0033-99EA 323 EUR
11-0033-99
Affinity chromatography columns, His GraviTrap™
Chromatographiesäulen
His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification.
  • For fast and simple purification of histidine-tagged proteins
  • Convenient, prepacked gravity-flow column
  • Allows direct purification of both clarified and unclarified cell lysates
  • High binding capacity; approximately 40 mg of pure protein per column
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives
The ready to use column is prepacked with Ni Sepharose 6 Fast Flow, which has a high protein binding capacity, low nickel-ion leakage, and is compatible with denaturing and reducing agents, as well a wide range of additives.
His GraviTrap columns are delivered in a package that can be converted into a column stand to simplify purification. LabMate PD-10 Buffer Reservoir can be connected to the columns for convenient handling of sample volumes above 10 ml.