Zinn ≥99,995% (Metall-Basis), Pulver -100 mesh

Lieferant: Thermo Fisher Scientific

11013.04 11013.09 11013.18 11013.30
11013.04EA 39.3 EUR
11013.04 11013.09 11013.18 11013.30
Zinn ≥99,995% (Metall-Basis), Pulver -100 mesh
Zinn

Purity: 99.995% CAS No.: 7440-31-5 Appearance: Grey Odour: Odourless MDL No.: MFCD00133862

Formel: Sn
Siedepunkt: 2270 °C (1013 hPa)
Schmelzpunkt: 232 °C
Dichte: 7,31 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00133862
CAS-Nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR