Iridium(III)chlorid Hydrat ≥99,8% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

11030.03 11030.06 11030.14 11030.18
11030.03EA 522 EUR
11030.03 11030.06 11030.14 11030.18
Iridium(III)chlorid Hydrat ≥99,8% (Metall-Basis)
Iridium(III)chlorid Hydrat

Purity: 99.9% CAS No.: 14996-61-3 Molecular Formula: IrCl3.xH2O Formula Weight: 298.56(anhy) Physical Form: Crystalline Appearance: Black Odour: Odourless Solubility: Soluble in water and alcohol Merck Reference: 14,5088 UN No.: 3260 MDL No.: MFCD00149750

Formel: Cl₃Ir·xH₂O
Dichte: 5,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149750
CAS-Nummer: 14996-61-3
EINECS: 233-044-6
UN: 3260
ADR: 8,II
Merck Index: 14,5088

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR