Hexachloroiridium(IV)säure (38 - 42% Ir) ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11031.03
11031.06
11031.14
11031.03EA
327
EUR
11031.03
11031.06
11031.14
Hexachloroiridium(IV)säure (38 - 42% Ir) ≥99% (Metall-Basis)
Hexachloroiridic(IV) acid hydrate
Purity: 99%
CAS No.: 110802-84-1
Molecular Formula: H2IrCl6.xH2O
Formula Weight: 406.93(anhy)
Physical Form: Lump
Appearance: Black
Solubility: Soluble in alcohol and water
UN No.: 3260
MDL No.: MFCD00011328
Formel:
H₂Cl₆Ir·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149906 CAS-Nummer: 110802-84-1 EINECS: 241-012-8 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...