Hexachloroplatinsäure Hydrat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.9%
CAS No.: 18497-13-7
Molecular Formula: H2PtCl6.6H2O
Formula Weight: 517.91 (409.82anhy)
Physical Form: Crystalline
Density: 2.430
Appearance: Red-brown
Odour: Odourless
Melting Point: 60° dec.
Solubility: Soluble in water, alcohol, acetone, ethyl acetate and ether
Merck Reference: 14,7526
UN No.: 2507
MDL No.: MFCD00149910
Catalysis, electroplating, photography, platinum mirrors, printing for etching of zinc, producing fine Colour in glass and porcelain, indelible ink, microscopy
Formel:
H₂Cl₆Pt·H₂O Dichte: 2,430 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149909 CAS-Nummer: 26023-84-7 EINECS: 607-848-6 UN: 2507 ADR: 8,III Merck Index: 14,7526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...