Cadmium(II)selenid ≥99,995% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cadmium (II) selenide

11057.18
11057.18EA 439 EUR
11057.18
Cadmium(II)selenid ≥99,995% (Metall-Basis)
Cadmium(II)selenid

Purity: 99.995% CAS No.: 1306-24-7 Molecular Formula: CdSe Formula Weight: 191.36 Physical Form: Powder Density: 5.81 Appearance: Brown-red to black Odour: Odourless Melting Point: 1350° Merck Reference: 14,1626 UN No.: 3077 MDL No.: MFCD00010917

Formel: CdSe
Molecular Weight: 191,37 g/mol
Schmelzpunkt: >1350 °C
Dichte: 5,81
Lagertemperatur: Raumtemperatur
MDL: MFCD00010917
CAS-Nummer: 1306-24-7
EINECS: 215-148-3
UN: 3283
ADR: 6.1,III
Merck Index: 14,1626

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR