Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 1.4 mm (0.055 in)

Lieferant: Thermo Fisher Scientific

Synonyme: Aurum

11063.BP 11063.BS 11063.C0
11063.BPEA 0 EUR
11063.BP 11063.BS 11063.C0
Gold ≥99,999% (Metall-Basis), Drahtform, Premion®, Ø 1.4 mm (0.055 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈29.71g/m Appearance: Gold Odour: Odourless MDL No.: MFCD00003436

Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Dichte: 19.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR