Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), Pulver -140 mesh Type 303-L
Lieferant: Thermo Scientific
|
Gefahr
|
11084.A1
11084.A1EA
359
EUR
11084.A1
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), Pulver -140 mesh Type 303-L
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:17:13 wt%
MDL No.: MFCD00148499