Vanadium(V)oxid ≥99,2%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.2%
CAS No.: 1314-62-1
Molecular Formula: V2O5
Formula Weight: 181.88
Physical Form: Powder
Density: 3.357
Melting Point: 690°
Boiling Point: 1750° dec.
Solubility: Slightly soluble in water. Soluble in acids and alkalies
Merck Reference: 14,9921
UN No.: 2862
MDL No.: MFCD00011457
As an oxidation catalyst, in refractories, as a ceramics Colourant, in the manufacture of vanadium salts, and as a dye
Formel:
O₅V₂ Siedepunkt: 1750 °C (Zers.) Schmelzpunkt: 690 °C Dichte: 3,357 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011457 CAS-Nummer: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III Merck Index: 14,9921 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...