Überbrille, Serie OX™
Lieferant: 3M
OX3000B 17-5118-3040P
111-1633EA 18.6 EUR
111-1633 MMMA17-5118-3040P
Überbrille, Serie OX™
Schutzbrillen
The 3M™ OX cover glasses (OTG) variants offer unique distinctive features: The OX2000 comes with adjustable lock-in temples (5 positions) and rotating tips (excellent grip in all positions) and the OX3000 is suitable for use with earmuffs; with low profile adjustable temples and non-slip pliable temple tips.
  • Optical Class 1 lens suitable for prolonged use
  • Protects against high speed particles at low energy (F) at extreme temperature conditions –5 and +55 °C (T)
  • Offers excellent protection against UV radiation
  • Designed to be worn over prescription eyewear
Can be used in a wide range of applications including construction, engineering, general assembly, light duty maintenance and repair and woodworking.
Standards: EN 166:2001, EN 170:2002