Wiederverwendbares Atemschutzgerät mit Halbmaske, HF-800 Serie, Secure Click™
Lieferant: 3M
HF-801 HF-803
111-2452EA 73.8 EUR
111-2452 111-2454
Wiederverwendbares Atemschutzgerät mit Halbmaske, HF-800 Serie, Secure Click™
Atemschutzmasken Mehrweg-Atemschutzmasken Halbmasken
Designed with smart and intuitive features, the 3M™ Secure Click™ half-mask reusable respirator is simple, comfortable and reliable. Available in three sizes: Small, medium and large.
  • Greater breathability and comfort provided by the world's first quad-flow cartridge system
  • Push-button seal check helps bolster confidence that the respirator is properly sealed to the face
  • New Secure Click™ connection ensures filters and cartridges are installed properly
  • Low profile half-mask design offers a wide field of view, and compatibility with certain welding and
More Product Information