Safety spectacles, Contemporary 571 Diamond, large
Lieferant: UNIVET
571030500
111-2455EA 23.9 EUR
111-2455
Safety spectacles, Contemporary 571 Diamond, large
Schutzbrillen
Contemporary series combines elegance and style in proetction, changing the concept of safety glasses.
  • Light and extremely tough
  • Integrated side shields
  • PA12 nylon frame with zero metallic components
  • Glossy varnish with shaded effect
  • Clear KN lenses, with anti-fog and anti-scratch ultra performing Vanguard PLUS treatment
Elegant and graceful for women, dynamic for men. Glasses can be employed for prolonged use as they do not create distorsions and allow a correct color perception, sensibly reducing visual fatigue.
Suitable for mechanical work in good visibility conditions.
Size: 56
Standards: EN-166, EN-170.