Face shield, disposable, 9710

Lieferant: UVEX
9710.514
111-2486EA 0 EUR
111-2486
Face shield, disposable, 9710
Gesichtsschutz
Designed specifically for use by healthcare professionals, this disposable face shield protects against drops and liquid splashes, reducing the risk of infections caused by droplets.

  • Protective face shield for single use
  • PET lens, 0,3 mm, with interior fog-free coating
  • Soft foam inner padding ensures comfortable wear, even for extended periods of use
  • One size fits all head sizes in the range 54 to 64 cm
  • Not suitable for protection against mechanical hazards, such as flying particles with higher energy
  • Space-saving flat-fold packaging

Due to its design, complete protection against viruses, bacteria, or other biological agents is only given to a limited extent. This face shield is only to be used by medical personnel or rescue workers and only for the duration of the current health threat. It can be combined with face protection masks, although it should be disposed of after a single use.

Standards: EN 166 PPE category II CE
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR