P3 R Hartschalen-Partikelfilter, Secure Click™
Lieferant: 3M
D9035
111-2570EA 456 EUR
111-2570
P3 R Hartschalen-Partikelfilter, Secure Click™
Atemschutzmasken Mehrweg-Atemschutzmasken Halbmasken
Secure Click™ P3 R D9035 hard case particulate filters are approved for use with 3M™ Secure Click™ respirators to provide protection against exposure to particles, dusts and mist in environments containing certain particles.
  • For use with 3M™ Secure Click™ respirators
  • Filter encapsulated in lightweight plastic case
  • Hard case recommended for harsh environments with a need for easy decontamination and protection
  • Hard case helps protect against sparks and high heat
  • Filter case designed for certain spray applications
  • Secure Click™ connection allows filters to be easily and securely inserted into place
  • Solid top and channelled airflow help protect filter
  • Swept-back design provides wide field of view
Get effective respiratory protection against exposure to certain particles, dusts and mists with 3M™ Secure Click™ P3 R D9035 hard case particulate filters. The P3 R filter is designed to protect against a wide range of particulate contaminates in higher heat applications. An innovative solid top air inlet helps reduce premature loading by allowing a channelled airflow. The comfortable, lightweight design provides a wide field of vision, and the filter can be clicked into place quickly and easily onto 3M™ Secure Click™ reusable respirators.
This series introduces an alternative to the 3M round bayonet connector with the new Secure Click™ connection, resulting in a quick, simple filter connection. It works just like a seat belt: Simply align the connections, push until audible click, and feel confident that filters are installed properly.