Safety spectacles, Rigi™
Lieferant: JSP
1RIG23CKN 1RIGBB21AR 1RIGBB21LO 1RIGLABB21AR 1RIG21C 1RIG23IO
JSPA1RIG23CKNEA 12.5 EUR
JSPA1RIG23CKN 111-2579 JSPA1RIGBB21LO 111-2580 JSPA1RIG21C JSPA1RIG23IO
Safety spectacles, Rigi™
Schutzbrillen
Ultra-lightweight safety spectacle with a wide field of vision to provide optimal coverage.
  • Large field of vsion
  • Wide range of lenses available
  • Optimal coverage
Whatever the workplace environment, Rigi™ spectacles deliver the perfect combination of performance, safety, and active style. The unique close fitting design, and ultra light weight (19g) ensure lasting comfort and fit. With a wide field of vision and a design to fit all faces, wearers can achieve a comfortable customised fit throughout the day. The ergonomic fit with flat and thin flexofitMore Product Information