Respirator filters for polimask 230
Lieferant: KLEIN ERWIN
422345 422346
111-3316EA 105 EUR
111-3316 KLNE422346
Respirator filters for polimask 230
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
The combined filters protect against organic gas, vapour and highly toxic compounds and the multi-type combined filter with additional protection against sulphur-di-oxide and ammonia. Suitable for Polimask 230.
  • Dolomite clogging test
Standards: EN 14387