Zirkonium(IV)oxid ≥99,978% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

11141.14 11141.22 11141.36
11141.14EA 238 EUR
11141.14 11141.22 11141.36
Zirkonium(IV)oxid ≥99,978% (Metall-Basis), Puratronic®
Zirkonium(IV)oxid

Purity: 99.978% CAS No.: 1314-23-4 Molecular Formula: O2Zr Formula Weight: 123.22 Physical Form: Powder Density: 5.89 Appearance: White Odour: Odourless Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310

Formel: ZrO₂
Siedepunkt: ∼5000 °C
Schmelzpunkt: ∼2700 °C
Dichte: 5,89
Lagertemperatur: Raumtemperatur
MDL: MFCD00011310
CAS-Nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR