Gallium(III)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11150.14
11150.22
11150.36
11150.06
11150.14EA
340
EUR
11150.14
11150.22
11150.36
11150.06
Gallium(III)nitrat Hydrat ≥99,999% (Metall-Basis), Puratronic®
Gallium(III)nitrat Hydrat
Purity: 99.999%
CAS No.: 69365-72-6
Molecular Formula: Ga(NO3)3.xH2O
Formula Weight: 255.73(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 110° dec.
Merck Reference: 14,4346
UN No.: 1477
MDL No.: MFCD00149723
Formel:
GaN₃O₉·H₂O Molecular Weight: 273.75 g/mol Schmelzpunkt: 110 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149723 CAS-Nummer: 69365-72-6 EINECS: 236-815-5 UN: 1477 ADR: 5.1 III Merck Index: 14,4346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...