Gallium(III) sulphate hydrate ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Purity: 99.999%
CAS No.: 13780-42-2
Molecular Formula: Ga2(SO4)3.xH2O (x≈18)
Formula Weight: 427.62(anhy)
Physical Form: Crystalline Aggregates
Density: 3.860
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 105-110°
MDL No.: MFCD00149725
Formel:
Ga₂(SO₄)₃·xH₂O (x≈18) Schmelzpunkt: 105…110 °C Dichte: 3,860 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149725 CAS-Nummer: 13780-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...