di-Bortrioxid, getrocknet (max. 0,02% H₂O) ≥99,999% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11160.QH
11160.QHEA
1121
EUR
11160.QH
di-Bortrioxid, getrocknet (max. 0,02% H₂O) ≥99,999% (Metall-Basis), Puratronic®
di-Bortrioxid
Purity: 99.999%
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Physical Form: Cylinder, 5.0cm dia, 55g/cylinder
Density: 2.46 +/- 0.01
Appearance: White
Odour: Odourless
Melting Point: ca 450°
Boiling Point: 1860°
Merck Reference: 14,1337
MDL No.: MFCD00011315
In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Formel:
B₂O₃ Siedepunkt: 1860 °C Schmelzpunkt: ∼450 °C Dichte: 2,46 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011315 CAS-Nummer: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...