Yttrium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Fisher Scientific
Achtung

11180.18 11180.30 11180.A1
11180.18EA 56.9 EUR
11180.18 11180.30 11180.A1
Yttrium(III)oxid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Yttrium(III)oxid

Purity: 99.9% CAS No.: 1314-36-9 Molecular Formula: Y2O3 Formula Weight: 225.81 Physical Form: 10 Micron Powder Density: 5.01 Appearance: White Odour: Odourless Melting Point: 2410° Solubility: Insoluble in water. Soluble in dilute acids Merck Reference: 14,10107 MDL No.: MFCD00011473


In phosphors for Colour TV tubes, in microwave filters, and as a stabilizer for high-temperature service materials

Formel: Y₂O₃
Siedepunkt: 4300 °C (1013 hPa)
Schmelzpunkt: 2410 °C
Dichte: 5,01
Lagertemperatur: Raumtemperatur
MDL: MFCD00011473
CAS-Nummer: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR