Yttrium(III)nitrat Hydrat ≥99,99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Yttrium trinitrate hydrate
11187.06
11187.14
11187.22
11187.59
11187.06EA
41.7
EUR
11187.06
11187.14
11187.22
11187.59
Yttrium(III)nitrat Hydrat ≥99,99%
Yttrium(III)nitrat Hydrat
Purity: 99.99%
Molecular Formula: Y(NO3)3.xH2O
Formula Weight: 274.92(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,10107
MDL No.: MFCD00011469
Formel:
N₃O₉Y·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011469 CAS-Nummer: 140191-40-8 EINECS: 233-802-6 UN: 1477 ADR: 5.1,III Merck Index: 14,10107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...