Yttrium(III)nitrat Hydrat ≥99,99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Yttrium trinitrate hydrate

11187.06 11187.14 11187.22 11187.59
11187.06EA 41.7 EUR
11187.06 11187.14 11187.22 11187.59
Yttrium(III)nitrat Hydrat ≥99,99%
Yttrium(III)nitrat Hydrat

Purity: 99.99% Molecular Formula: Y(NO3)3.xH2O Formula Weight: 274.92(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,10107 MDL No.: MFCD00011469

Formel: N₃O₉Y·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00011469
CAS-Nummer: 140191-40-8
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III
Merck Index: 14,10107

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR