Histopaque®, Filter sterile, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung
11191-100ML 11191-6X100ML
11191-100MLEA 165 EUR
11191-100ML 11191-6X100ML
Histopaque®, Filter sterile, Sigma-Aldrich®
Histopaque®
Histopaque®-1119 is a sterile, endotoxin tested, ready-to-use medium. This solution consists of polysucrose and sodium diatrizoate, adjusted to a density of 1,119 g/ml and pH 8,8 to 9,0.
  • Histopaque®-1119 offers numerous advantages, such as:
  • Optimal recovery of viable cells
  • Permits selective separation of blood cell types
  • Prevents cell distortion
  • Preserves cell viability
  • Minimal extraneous cell interference
  • Reproducible performance from batch to batch
  • Sterile-filtered and endotoxin-tested
  • Stable at 2 to 8 °C for at least 2 years when protected from light
Histopaque®-1119 is suitable for use in conjunction with Histopaque-1077 as it facilitates the rapid recovery of viable mononuclear cells and granulocytes from small volumes of whole blood or bone marrow.