Heat resistant gloves
Lieferant: JUTEC
H3110040
112-1964EA 421 EUR
112-1964
Heat resistant gloves
Handschuhe
These soft HT-fabric mittens with thumb are ideal for use when working with hot glass up to 1100 °C.
  • Ambidextrous
  • Double insulated
  • Does not stick to glass
Typical applications include: Furnace construction, laboratory work, handling of hot glass.
Colour: Cream
EN 388 (1XX2), EN 407 (44X21X); Cat. III
Standards: EN 388 (1XX2), EN 407 (44X21X); Cat. III.