Nitrile gloves, Microflex® 93-823
Lieferant: Ansell
93-823/6.5-7 93-823/7.5-8
112-4322EA 0 EUR
112-4322 112-4323
Nitrile gloves, Microflex® 93-823
Handschuhe
These disposable nitrile gloves provide chemical and liquid protection. The most advanced glove in type I and IV allergy prevention, the Microflex 93-823 has been clinically proven to decrease redness and itchiness associated with contact dermatitis. Engineered without the use of chemical additives (including accelerators) that are known skin irritants, the 93-823 is safe for those with sensitive skin or prone to allergic reactions.
  • Non sterile, powder-free
  • Decreases skin irritation potential in sensitive hands
  • Textured fingertips and a reduced amount of surfactant used to produce the glove results in a secure and confident grip for handling objects
  • Increased sensitivity due to the thin polymer design
  • Ultra-soft materials provide comfort and exceptional fit for long wear periods
Ideal for use in analytical testing/measurements, assembly and inspection, biotechnologies, cleaning, janitorial and maintenance work, laboratory analysis, pharmaceutical production and light duty assembly.
Standards: EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 VIRUS; Cat. III; AQL 1,5