Reinraumhandschuhe, Nitril, steril, BioClean Emerald™
Lieferant: Nitritex
BENS8.0 BENS8.5 BENS10.0 BENS6.0 BENS6.5 BENS7.0 BENS7.5 BENS9.0
112-4959EA 616 EUR
112-4959 112-6364 112-4954 112-4955 112-4956 112-4957 112-4958 112-5111
Reinraumhandschuhe, Nitril, steril, BioClean Emerald™
Handschuhe
Hand-specific chemotherapy gloves with beaded cuffs. Suitable for double-donning and for handling cytotoxic drugs.
  • Sterile, powder-free, textured finish
  • Latex-free to eliminate Type 1 latex allergies
  • Hypoallergenic
  • Nitrile is resistant to a wide range of chemicals
  • Each size carries a unique bar code on the carton (ITF-14), which allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 455, EN 374-2; Cat. III; AQL 1,5
Standards: EN 455, EN ISO 374-1 Typ B, EN ISO 374-5 VIRUS, EN 421, EN 1149-5, Kat. III.
Für den Kontakt mit Lebensmitteln zugelassen.
Verpackung: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs).