Kaliumiodid ≥99,0%, AR® ACS, frei fließend, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Gefahr
|
Synonyme:
Iodkalium
Veränderungsmanagement-Kategorie = N
1123-20EA
0
EUR
1123-20
Kaliumiodid ≥99,0%, AR® ACS, frei fließend, Macron Fine Chemicals™
Kaliumiodid
Formel:
Kl Siedepunkt: 1330 °C (1013 hPa) Schmelzpunkt: 681 °C Dichte: 3,13 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011405 CAS-Nummer: 7681-11-0 EINECS: 231-659-4 Merck Index: 13,07727 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | ≥99.0% |
ACS - Assay (KI) | ≥99.0 % |
ACS - Appearance (white, crystalline granules) | Passes Test |
ACS - Barium (Ba) | ≤0.002 % |
ACS - Calcium (Ca) | ≤0.002 % |
ACS - Chloride and Bromide (as Cl) | ≤0.01 % |
ACS - Heavy Metals (as Pb) | ≤0.0005 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Iodate (IO₃) | ≤3 ppm |
ACS - Iron (Fe) | ≤0.0003 % |
ACS - Loss on Drying at 150°C | ≤0.2 % |
ACS - Magnesium (Mg) | ≤0.001 % |
ACS - pH of 5% Solution at 25°C | 6.0 - 9.2 |
ACS - Phosphate (PO₄) | ≤0.001 % |
ACS - Sodium (Na) | ≤0.005 % |
ACS - Sulfate (SO₄) | ≤0.005 % |
Free Alkali, For Information Only | For Information Only |
Total Alkali, For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...