Praseodymtrifluorid, wasserfrei ≥99,9895% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
11241.09
11241.18
11241.09EA
153
EUR
11241.09
11241.18
Praseodymtrifluorid, wasserfrei ≥99,9895% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Praseodymtrifluorid
Purity: 99.9895%
CAS No.: 13709-46-1
Molecular Formula: F3Pr
Formula Weight: 197.90
Physical Form: Powder
Density: 6.2
Appearance: Green
Odour: Odourless
Melting Point: 1395°
UN No.: 3288
MDL No.: MFCD00016287
Formel:
F₃Pr Schmelzpunkt: 1395 °C Dichte: 6,2 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016287 CAS-Nummer: 13709-46-1 EINECS: 237-254-9 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...