Biohazard bag holders, Sterilin®
Lieferant: Sterilin
S23B
113-0093EA 291 EUR
113-0093
Biohazard bag holders, Sterilin®
Gestelle Beutelständer
Coated wire or cardboard holders for use with autoclave bags.