Certriacetat Sesquihydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Essigsäure Cer(III)salz Sesquihydrat, Cer(III)acetat Sesquihydrat

11324.09 11324.18 11324.30
11324.09EA 53.3 EUR
11324.09 11324.18 11324.30
Certriacetat Sesquihydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Certriacetat Sesquihydrat

Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.


Purity: 99.99% CAS No.: 537-00-8 Molecular Formula: Ce(C2H3O2)3.1.5H2O Formula Weight: 344.28 (317.26anhy) Physical Form: Crystalline Aggregates Melting Point: 308° dec. MDL No.: MFCD00051060

Formel: Ce(C₂H₃O₂)₃·1.5H₂O
Schmelzpunkt: 308 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051060
CAS-Nummer: 537-00-8
EINECS: 208-654-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR