Certriacetat Sesquihydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Essigsäure Cer(III)salz Sesquihydrat, Cer(III)acetat Sesquihydrat
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.
Purity: 99.99%
CAS No.: 537-00-8
Molecular Formula: Ce(C2H3O2)3.1.5H2O
Formula Weight: 344.28 (317.26anhy)
Physical Form: Crystalline Aggregates
Melting Point: 308° dec.
MDL No.: MFCD00051060
Formel:
Ce(C₂H₃O₂)₃·1.5H₂O Schmelzpunkt: 308 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051060 CAS-Nummer: 537-00-8 EINECS: 208-654-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...