Ammoniaklösung 25%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

1133.1000EA 30.5 EUR
1133.1000 1133.2500 1133.9025
Ammoniaklösung 25%, AnalaR NORMAPUR® analytisches Reagens
Ammoniaklösung
Formel: NH₃
Siedepunkt: 32…37,7 °C (1013 hPa)
Schmelzpunkt: –60…–57,5 °C
Dichte: 0,907 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Assay Min. 25 %
Evaporation residue Max. 20 ppm
Substances reducing KMnO4 (as O) Max. 8 ppm
Total S (as SO4) Max. 1 ppm
Cl (Chloride) Max. 0.5 ppm
CO3 (as CO2) Max. 15 ppm
PO4 (Phosphate) Max. 1 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.1 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 1 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 2 ppm
Pb (Lead) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR