Cobalt(II)nitrat Hexahydrat ≥97,7%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Cobaltnitrat Hexahydrat, Cobaltdinitrat Hexahydrat
11341.14
11341.30
11341.A1
11341.A7
11341.14EA
59.2
EUR
11341.14
11341.30
11341.A1
11341.A7
Cobalt(II)nitrat Hexahydrat ≥97,7%
Cobalt(II)nitrat Hexahydrat
Purity: 97.7% min
CAS No.: 10026-22-9
Molecular Formula: Co(NO3)2.6H2O
Formula Weight: 291.04 (182.94anhy)
Physical Form: Crystalline
Density: 1.87
Refractive Index: 1.52
Appearance: Red
Odour: Odourless
Melting Point: 55-56°
Merck Reference: 14,2444
UN No.: 1477
MDL No.: MFCD00149647
Formel:
Co(NO₃)₂·6H₂O Schmelzpunkt: 55…56 °C Dichte: 1,87 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149647 CAS-Nummer: 10026-22-9 EINECS: 233-402-1 UN: 1477 ADR: 5.1,III Merck Index: 14,2444 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...