Organic reference standard, disperse yellow 3, analytical standard, Supelco®
Lieferant: Merck
Disperse Yellow 3 is an yellow azo dye which is majorly used in textile industry. It also has carcinogenic properties.
It is used to understand that Phanerochaete chrysosporium can mineralise azo dyes with radiolabelled substrate.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.