Lithium thiocyanate hydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
11349.18
11349.18EA
163
EUR
11349.18
Lithium thiocyanate hydrate
Lithium thiocyanate hydrate
CAS No.: 123333-85-7
Molecular Formula: LiSCN.xH2O
Formula Weight: 65.02(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00149773
Formel:
LiSCN·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149773 CAS-Nummer: 123333-85-7 EINECS: 209-135-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...