Silber ≥99,999% (Metall-Basis), Perlen, Premion® 1-7 mm (0.04-0.1 in)
Lieferant: Thermo Fisher Scientific
11357.09
11357.18
11357.30
11357.09EA
127
EUR
11357.09
11357.18
11357.30
Silber ≥99,999% (Metall-Basis), Perlen, Premion® 1-7 mm (0.04-0.1 in)
Silver
Purity: 99.999%
CAS No.: 7440-22-4
Odour: Characteristic
MDL No.: MFCD00003397
For vapor deposition
Formel:
Ag Siedepunkt: 2212 °C (1013 hPa) Schmelzpunkt: 962 °C Dichte: 10,5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003397 CAS-Nummer: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...