Gold ≥99,9975% (Metall-Basis), Folie, Premion®, Dicke 0.1 mm (0.004 in)

Lieferant: Thermo Fisher Scientific

Synonyme: Aurum

11391.FF 11391.FI 11391.GH
11391.FFEA 362 EUR
11391.FF 11391.FI 11391.GH
Gold ≥99,9975% (Metall-Basis), Folie, Premion®, Dicke 0.1 mm (0.004 in)
Gold

Purity: 99.9975+% CAS No.: 7440-57-5 Physical Form: ≈1.2g/25x25mm Odour: Odourless MDL No.: MFCD00003436

Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Dichte: 19.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR