Zirkonium(IV)oxid ≥99% Metall-Basis ausgeschlossen Hf (max. 2% HfO₂)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99+%
CAS No.: 1314-23-4
Molecular Formula: ZrO2
Formula Weight: 123.22
Physical Form: -325 Mesh Powder
Density: 5.89
Melting Point: ca 2700°
Boiling Point: ca 5000°
Solubility: Insoluble in water. Soluble in sulfuric acid and hydrofluoric acid. Dissolves on heating in a mixture of 2 parts H2SO4 and 1 part water
Merck Reference: 14,10180
MDL No.: MFCD00011310
Unstabilized: In production of piezoelectric crystals, in high-frequency induction coils, and in ceramic glazes. Stabilized: For refractory furnace linings, and in crucibles
Formel:
ZrO₂ Siedepunkt: ∼5000 °C Schmelzpunkt: ∼2700 °C Dichte: 5,89 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011310 CAS-Nummer: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...