Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, Renzo Biomex
Lieferant: ELTEN
763421/36 763421/37 763421/38 763421/39 763421/40 763421/41 763421/42 763421/43 763421/44 763421/45 763421/46 763421/47 763421/48 763421/49 763421/50
114-7798EA 115.9 EUR
114-7798 114-7799 114-7800 114-7801 114-7802 114-7803 114-7804 114-7805 114-7806 114-7807 114-7808 114-7809 114-7810 114-7811 114-7812
Halbhohe ESD-Sicherheitsschuhe, zum Schnüren, Renzo Biomex
Schuhe und Stiefel
Durable ESD safety shoes made from robust, breathable cowhide leather and hydrophobised nubuck leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Anatomically-shaped insole and ankle support for improved stability
  • Biomex-Protection®: Cuff stabilises ankles
  • Heat resistant seams and laces
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black.
Standards: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, CI.