Disposable conductive booties
Lieferant: DESCO
52016
114-9321EA 223 EUR
114-9321
Disposable conductive booties
Einweg-Kleidung
Disposable conductive surgical style Booties are 'one size fits all' with elastic opening.
  • Lightweight
  • Cleanroom compatible
  • Maintains grounding with full mobility
  • Will not scratch or mark flooring
Disposable ESD Booties can be used for person/footwear/flooring system per EN 61340-5-1.
Verpackung: Sold 150 pairs per box.