Tychem® 6000 AL TF640 00 Ventilated Coveralls

Lieferant: DuPont
TF0640TGY00/S TF0640TGY00/3X TF0640TGY00/L TF0640TGY00/M TF0640TGY00/XL TF0640TGY00/2X
114-9615EA 485 EUR
114-9615 114-9620 114-9617 114-9616 114-9618 114-9619
Tychem® 6000 AL TF640 00 Ventilated Coveralls
Einweg-Kleidung
Air-fed coverall with cuff elastication and over boot assembly, consisting of integrated dissipative inner boots attached to dissipative outer boots.

  • Ventilated coverall with stitched and overtaped seams
  • Fully enclosed over hood with clear front and crown see-through visor
  • Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
  • Three exhaust valves; two at hips, one in hood back
  • Air distribution made of Tychem 6000 fabric
  • Emergency egress
  • Cuff elastication and elastic thumb loop on inner sleeve opening
  • Over boot assembly consists of integrated dissipative inner boots attached to dissipative outer boots with buckles

Tychem® 6000 AL garments are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to Tyvek® substrate. They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards, and certain chemical warfare agents.

Tychem® 6000 AL garments are designed to help keep workers safe and focused when carrying out critical tasks in industries like Pharmaceutics, chemicals and others.

Colour: Grey

Cuff elastication and elastic thumb loop on inner sleeve opening.
Over boot assembly consisting of integrated dissipative inner boots attached to dissipative outer boots with buckles.
Simplified closure: easier donning procedure, minimal help from an assistant required.
Airline belt inside the suit: high reusability of the belt/air-regulator.
Visor with 180° panoramic view: clear vision, non-PVC, highly chemical resistant solution.
Top visor: in suit lighting and vision above suit; improved safety with secured sights and comfort.
Emergency egress strip: quick doffing in case of emergency (tear off at once).
Fabric air plenum distributing cooling and breathing air for optimal comfort and reduced heat stress.
Simple breathing airline belt/regulator interface and installation: easy and quick installation set-up, no taping required.

Standards: Certified according to Regulation (EU) 2016/425
Chemical protective clothing, Category III, Type 3-B, 4-B and 6-B
EN 14126 (barrier to infective agents)
EN 1073-1 (ventilated protective clothing against particulate radioactive contamination)
Antistatic treatment (EN 1149-5)
Certified system made of Tychem® 6000 AL TF64000 and DuPont air-belt, model 8000513914, to protect the worker according to EN 14594 (Respiratory protective devices - Continuous flow compressed air line breathing devices)

Verpackung: Two per box, individually packed.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR