Iridium ≥99,8% (Metall-Basis), Drahtform, Ø 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific

11429.BO 11429.BQ
11429.BOEA 338 EUR
11429.BO 11429.BQ
Iridium ≥99,8% (Metall-Basis), Drahtform, Ø 1.0 mm (0.04 in)
Iridium

Purity: 99.8% CAS No.: 7439-88-5 Physical Form: ≈17.8g/m MDL No.: MFCD00011062

Formel: Ir
Siedepunkt: 5300 °C (1013 hPa)
Schmelzpunkt: 2450 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011062
CAS-Nummer: 7439-88-5
EINECS: 231-095-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR