Bromkresolgrün Natriumsalz in ACS analysereinem Wasser ≥90% (durch HPLC), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 3,3',5,5'-Tetrabrom-m-kresolsulfonphthalein Natriumsalz, Bromkresolgrün wasserlöslich

M-Clarity™ Qualitätsstufe = MQ200

114367-25G 114367-5G
114367-25GEA 285 EUR
114367-25G 114367-5G
Bromkresolgrün Natriumsalz in ACS analysereinem Wasser ≥90% (durch HPLC), Sigma-Aldrich®
Bromkresolgrün Natriumsalz

Bromocresol Green (BCG), also known as bromcresol green, is a synthetic, anionic, triphenylmethane dye belonging to the phthalein and sulphonphthalein dyes group. It is commonly used as a pH indicator as it changes colour from yellow to blue-green at pH 3,8 to 5,4. The monosodium salt of bromocresol green is highly ionisable in water. The dye at a higher pH level produces a large and weakly lipophilic dianion.


  • Bromocresol green serves as a pH indicator in biology and medicinal experiments
  • It is used widely as a vital stain to study blood-brain permeability
  • It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations, and in charge-transfer complexation processes
  • It is used in TLC for the visualisation of compounds with functional groups whose pKa is below 5
  • It has been used in the pH-sensitive colorimetric determination of glutamate decarboxylase activity
  • It has also been employed in the evaluation of paper-analytical devices for colorimetric analysis of serum samples
  • It is suitable as a selective inhibitor in kidney physiology studies
  • It is a diagnostic assay manufacturing, hematology, histology

Formel: C₂₁H₁₃Br₄NaO₅S
Schmelzpunkt: 230 °C
Lagertemperatur: Room Temperature
MDL: MFCD00148898
CAS-Nummer: 62625-32-5
EINECS: 263-657-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6576f0c57895b913ac2b0cdaad5ae705df1a91ad&vid=e5e334727f1a16bf4bbcf034a9586a032e897bd9&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Visual Interval 1, pH 3.8 = Yellow Pass
Visual Interval 2, pH 4.5 = Green Pass
Visual Interval 3, pH 5.4 = Blue Pass
Clarity of solution not more than a faint trace of turbidity or insoluble matter c = 0.1 g/100 ml; Water Conforms
Purity (HPLC) ≥90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR