Bromthymolblau Natriumsalz in ACS analysereinem Wasser, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

114421-25G 114421-5G
114421-25GEA 123 EUR
114421-25G 114421-5G
Bromthymolblau Natriumsalz in ACS analysereinem Wasser, Sigma-Aldrich®
Bromthymolblau Natriumsalz

Bromothymol blue has been used for freshwater pH measurements over a broad range of buffer intensities.


  • pH-indicator
  • Used for carbonic anhydrase assay due to the conversion of carbon dioxide to bicarbonate
  • Used as a diagnostic assay manufacturing, hematology, histology

Formel: C₂₇H₂₇Br₂NaO₅S
Schmelzpunkt: >200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00077263
CAS-Nummer: 34722-90-2
EINECS: 252-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f354f58906e22f2bb240fbb7582547d19079330&vid=5b5d658a2b6164adbb36ca73400e0f81bd70054a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Wavelength c = 0.005 g/L; 0.1 N NaOH 614 - 618 nm
Extinction Coefficient ≥13000
Wavelength 305 - 309 nm
Visual Transition Interval Yellow to Yellow-Green (pH 6.0) Pass
Visual Transition Interval Green (pH 6.7) Pass
Visual Transition Interval Blue (pH 7.6) Pass
Clarity of Solution, c = 0.1%; Water, Clear with only a faint trace of turbidity or insoluble matter Clear to Very Slightly Hazy


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR