Eisen ≥99,995% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific

11448.FF 11448.FI
11448.FFEA 220 EUR
11448.FF 11448.FI
Eisen ≥99,995% (Metall-Basis), Folie, Puratronic®, Dicke 1.0 mm (0.04 in)
Iron

Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈4.9g/25x25mm MDL No.: MFCD00010999

Formel: Fe
Siedepunkt: 3000 °C (1013 hPa)
Schmelzpunkt: 1535 °C
Dichte: 7,86 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010999
CAS-Nummer: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR