Methylorange water_ACS reagent grade 85% Dye, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Goldorange, Helianthin, Natrium-4-(4-dimethylaminophenylazo)benzolsulfonat, Orange III, 4-(4-(Dimethylamino)phenylazo)benzolsulfonsäure Natriumsalz, (4-(Dimethylamino)azobenzol-4'-sulfonsäure Natriumsalz

MQ200

114510-25G 114510-100G 114510-500G
114510-25GEA 36 EUR
114510-25G 114510-100G 114510-500G
Methylorange water_ACS reagent grade 85% Dye, Sigma-Aldrich®
Methylorange

Methyl orange (C.I. 13025) is the sodium salt of p-dimethylaminoazobenzenesulfonic acid. It is an orange-colored synthetic dye, which changes color depending on the pH of the solution. It develops a red color in acidic solution and yellow color in basic solution.


  • Methyl orange is used as a:
  • pH indicator in the cell sap
  • Counterstain to crystal violet in staining pollen tubes
  • Component of a polychrome histological stain
  • Solution indicator, such as in assays of oxalic and malonic acids
  • In other biomedical applications that include use in film dosimeters and as a reagent for the assay of bromide ions

Formel: C₁₄H₁₄N₃NaO₃S
Molecular Weight: 327.34 g/mol
Siedepunkt: ∼90 °C (1013 hPa)
Schmelzpunkt: >300 °C
Dichte: ∼1 g/cm³ (20 °C)
Flammpunkt: 38
Lagertemperatur: Raumtemperatur
MDL: MFCD00007502
CAS-Nummer: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1 III
Merck Index: 13,06127
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=915f043445c2be992ab36bc290d60762365a46ad&vid=bd67cfd7651199e11bfc8312958675f5db23665e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Orange
Appearance (form) Powder
Infrared spectrum Conforms to structure
Visual interval, pH 3.0 pink or red Pass
Visual interval, pH 3.2 pinkish orange Pass
Visual interval, pH 4.4 yellow Pass
Appearance (clarity) Pass
Carbon 43.7 - 59.1%
Nitrogen 10.9 - 14.8%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR